ADVH49B830C0 AChemBlock

3-hydroxy-2,3-dimethylbutan-2-yl hydrogen (3-methoxyprop-1-yn-1-yl)boronate

  • Cas Number : 634196-63-7
  • Molecular Weight : 214.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-hydroxy-2-3-dimethylbutan-2-yl-hydrogen-3-methoxyprop-1-yn-1-yl-boronate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72329
Availability details for
Quantity
  • Availability
  • Add to Cart