ADVH49B830C2 AChemBlock

1M7

  • Cas Number : 73043-80-8
  • Molecular Weight : 222.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/1m7.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86684
Availability details for
Quantity
  • Availability
  • Add to Cart