ADVH49B830C7 AChemBlock

CMP8

  • Cas Number : 851107-28-3
  • Molecular Weight : 528.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/cmp8.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86596
Availability details for
Quantity
  • Availability
  • Add to Cart