ADVH49B830CC AChemBlock

methyl (3r,5r)-1-oxaspiro[2.3]hexane-5-carboxylate

  • Cas Number : 578715-80-7
  • Molecular Weight : 142.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-3r-5r-1-oxaspiro-2-3-hexane-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S74958
Availability details for
Quantity
  • Availability
  • Add to Cart