ADVH49B830CD AChemBlock

3-(aminomethyl)tetrahydrothiophene 1,1-dioxide hydrochloride

  • Cas Number : 3193-51-9
  • Molecular Weight : 185.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-aminomethyl-tetrahydrothiophene-1-1-dioxide-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76128
Availability details for
Quantity
  • Availability
  • Add to Cart