ADVH49B830D3 AChemBlock

7-(tert-butyl) 3-ethyl 1-oxa-2,7-diazaspiro[4.4]non-2-ene-3,7-dicarboxylate

  • Cas Number : 1824081-61-9
  • Molecular Weight : 298.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-tert-butyl-3-ethyl-1-oxa-2-7-diazaspiro-4-4-non-2-ene-3-7-dicarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78042
Availability details for
Quantity
  • Availability
  • Add to Cart