ADVH49B830D5 AChemBlock

[3-[[tert-butyl(dimethyl)silyl]oxymethyl]-1,2,3,5,6,7-hexahydropyrrolizin-8-yl]methanol

  • Cas Number : 1788873-94-8
  • Molecular Weight : 285.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-tert-butyl-dimethyl-silyl-oxymethyl-1-2-3-5-6-7-hexahydropyrrolizin-8-yl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78300
Availability details for
Quantity
  • Availability
  • Add to Cart