ADVH49B830D6 AChemBlock

ethyl 6,6-difluoro-3-oxo-1,2,5,7-tetrahydropyrrolizine-8-carboxylate

  • Cas Number : 1788873-47-1
  • Molecular Weight : 233.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-6-6-difluoro-3-oxo-1-2-5-7-tetrahydropyrrolizine-8-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78301
Availability details for
Quantity
  • Availability
  • Add to Cart