ADVH49B830D7 AChemBlock

dichlororuthenium;2-(2-pyridyl)pyridine;dihydrate

  • Cas Number : 15746-57-3
  • Molecular Weight : 520.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/dichlororuthenium-2-2-pyridyl-pyridine-dihydrate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79235
Availability details for
Quantity
  • Availability
  • Add to Cart