ADVH49B830D9 AChemBlock

4-chloro-2-(7,7-dimethyl-1-oxo-1,3,4,6,7,8-hexahydro-2H-cyclopenta[4,5]pyrrolo[1,2-a]pyrazin-2-yl)nicotinaldehyde

  • Cas Number : 1434050-55-1
  • Molecular Weight : 343.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-2-7-7-dimethyl-1-oxo-1-3-4-6-7-8-hexahydro-2h-cyclopenta-4-5-pyrrolo-1-2-a-pyrazin-2-yl-nicotinaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S79944
Availability details for
Quantity
  • Availability
  • Add to Cart