ADVH49B830DA AChemBlock

6-thia-1-azaspiro[3.3]heptane 6,6-dioxide oxalate

  • Cas Number : 1392804-12-4
  • Molecular Weight : 237.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-thia-1-azaspiro-3-3-heptane-6-6-dioxide-oxalate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S80287
Availability details for
Quantity
  • Availability
  • Add to Cart