ADVH49B830DB AChemBlock

sodium 5-oxo-5H-pyrazolo[1,5-a]pyrimidin-4-ide

  • Cas Number : 1224944-43-7
  • Molecular Weight : 157.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/sodium-5-oxo-5h-pyrazolo-1-5-a-pyrimidin-4-ide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82194
Availability details for
Quantity
  • Availability
  • Add to Cart