ADVH49B830DD AChemBlock

(1R,2S)-2-(aminomethyl)cyclohexan-1-ol

  • Molecular Weight : 129.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-2s-2-aminomethyl-cyclohexan-1-ol-307624.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84314
Availability details for
Quantity
  • Availability
  • Add to Cart