ADVH49B830DE AChemBlock

3-amino-5-methyl-1H-pyrazole-4-carbonitrile

  • Molecular Weight : 122.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-amino-5-methyl-1h-pyrazole-4-carbonitrile-307627.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84317
Availability details for
Quantity
  • Availability
  • Add to Cart