ADVH49B830E0 AChemBlock

trans-tert-butyl N-4-(propan-2-yl)pyrrolidin-3-yl]carbamate

  • Molecular Weight : 228.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/trans-tert-butyl-n-4-propan-2-yl-pyrrolidin-3-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84346
Availability details for
Quantity
  • Availability
  • Add to Cart