ADVH49B830E1 AChemBlock

tert-butyl ((1S,3S)-3-cyanocyclopentyl)carbamate

  • Molecular Weight : 210.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1s-3s-3-cyanocyclopentyl-carbamate-307657.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84347
Availability details for
Quantity
  • Availability
  • Add to Cart