ADVH49B830E6 AChemBlock

tert-butyl N-[cis-3-methoxypiperidin-4-yl]carbamate hemioxalate

  • Molecular Weight : 550.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-cis-3-methoxypiperidin-4-yl-carbamate-hemioxalate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84427
Availability details for
Quantity
  • Availability
  • Add to Cart