ADVH49B830EA AChemBlock

tert-butyl 3-oxa-2-thia-1-azaspiro[4.4]nonane-1-carboxylate 2,2-dioxide

  • Molecular Weight : 277.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-oxa-2-thia-1-azaspiro-4-4-nonane-1-carboxylate-2-2-dioxide-307994.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84684
Availability details for
Quantity
  • Availability
  • Add to Cart