ADVH49B830EB AChemBlock

tert-butyl 1-oxospiro[indane-2,3'-piperidine]-1'-carboxylate

  • Molecular Weight : 301.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1-oxospiro-indane-2-3-apos-piperidine-1-apos-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84690
Availability details for
Quantity
  • Availability
  • Add to Cart