ADVH49B830EC AChemBlock

tert-butyl (7R)-7-methyl-1,4-diazepane-1-carboxylate;(2S,3S)-2,3-dibenzoyloxybutanedioic acid

  • Molecular Weight : 786.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-7r-7-methyl-1-4-diazepane-1-carboxylate-2s-3s-2-3-dibenzoyloxybutanedioic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84708
Availability details for
Quantity
  • Availability
  • Add to Cart