ADVH49B830EF AChemBlock

NTRC-0066-0

  • Cas Number : 1817791-73-3
  • Molecular Weight : 565.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ntrc-0066-0.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S78164
Availability details for
Quantity
  • Availability
  • Add to Cart