ADVH49B830F0 AChemBlock

spiro[1,10-diazatricyclo[6.4.0.02,6]dodeca-2(6),7-diene-5,1'-cyclopropane]-9-one

  • Molecular Weight : 202.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/spiro-1-10-diazatricyclo-6-4-0-0-sup-2-6-sup-dodeca-2-6-7-diene-5-1-apos-cyclopropane-9-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84830
Availability details for
Quantity
  • Availability
  • Add to Cart