ADVH49B830F4 AChemBlock

methyl cis-5-methylpiperidine-3-carboxylate hemioxalate

  • Molecular Weight : 404.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-cis-5-methylpiperidine-3-carboxylate-hemioxalate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84880
Availability details for
Quantity
  • Availability
  • Add to Cart