ADVH49B830F8 AChemBlock

(1R,5S,6s)-3-thiabicyclo[3.1.0]hexane-6-carboxylic acid 3,3-dioxide

  • Molecular Weight : 176.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-5s-6s-3-thiabicyclo-3-1-0-hexane-6-carboxylic-acid-3-3-dioxide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85010
Availability details for
Quantity
  • Availability
  • Add to Cart