ADVH49B830FC AChemBlock

8-thia-2-azaspiro[4.5]decane 8,8-dioxide hydrochloride

  • Molecular Weight : 225.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/8-thia-2-azaspiro-4-5-decane-8-8-dioxide-hydrochloride-308469.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85159
Availability details for
Quantity
  • Availability
  • Add to Cart