ADVH49B83100 AChemBlock

6-thia-2-azaspiro[3.4]octane 6,6-dioxide hydrochloride

  • Molecular Weight : 197.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-thia-2-azaspiro-3-4-octane-6-6-dioxide-hydrochloride-308529.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85219
Availability details for
Quantity
  • Availability
  • Add to Cart