ADVH49B83103 AChemBlock

6-hydroxy-1-thiaspiro[3.3]heptane 1,1-dioxide

  • Molecular Weight : 162.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-hydroxy-1-thiaspiro-3-3-heptane-1-1-dioxide-308544.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85234
Availability details for
Quantity
  • Availability
  • Add to Cart