ADVH49B83108 AChemBlock

5-methylspiro[indoline-3,4'-piperidine]-2-one;hydrochloride

  • Molecular Weight : 252.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-methylspiro-indoline-3-4-apos-piperidine-2-one-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S85290
Availability details for
Quantity
  • Availability
  • Add to Cart