ADVH49B83109 AChemBlock

DAAO inhibitor-1

  • Cas Number : 2065250-25-9
  • Molecular Weight : 152.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/daao-inhibitor-1.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86770
Availability details for
Quantity
  • Availability
  • Add to Cart