ADVH49B8310D AChemBlock

WYC-209

  • Cas Number : 2131803-90-0
  • Molecular Weight : 368.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/wyc-209.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86690
Availability details for
Quantity
  • Availability
  • Add to Cart