ADVH49B8311C AChemBlock

(R)-2-((tert-butoxycarbonyl)amino)-2-(1,1-dioxidotetrahydro-2H-thiopyran-4-yl)acetic acid

  • Molecular Weight : 307.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/r-2-tert-butoxycarbonyl-amino-2-1-1-dioxidotetrahydro-2h-thiopyran-4-yl-acetic-acid-309413.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86103
Availability details for
Quantity
  • Availability
  • Add to Cart