ADVH49B8311E AChemBlock

(1R,5S,9s)-9-hydroxy-9-methyl-3-thia-7-azabicyclo[3.3.1]nonane 3,3-dioxide

  • Molecular Weight : 205.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-5s-9s-9-hydroxy-9-methyl-3-thia-7-azabicyclo-3-3-1-nonane-3-3-dioxide-309474.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86164
Availability details for
Quantity
  • Availability
  • Add to Cart