ADVH49B83121 AChemBlock

(1R,5S)-7-benzyl-3-thia-7-azabicyclo[3.3.1]nonan-9-one 3,3-dioxide

  • Molecular Weight : 279.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-5s-7-benzyl-3-thia-7-azabicyclo-3-3-1-nonan-9-one-3-3-dioxide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86168
Availability details for
Quantity
  • Availability
  • Add to Cart