ADVH49B83124 AChemBlock

Ethyl 5-(tert-butyl)-1,2,4-oxadiazole-3-carboxylate

  • Molecular Weight : 198.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/ethyl-5-tert-butyl-1-2-4-oxadiazole-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=K16163
Availability details for
Quantity
  • Availability
  • Add to Cart