ADVH49B83125 AChemBlock

YKL-06-061

  • Cas Number : 2172617-15-9
  • Molecular Weight : 527.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/ykl-06-061.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86602
Availability details for
Quantity
  • Availability
  • Add to Cart