ADVH49B83126 AChemBlock

ZK-811752

  • Cas Number : 217645-70-0
  • Molecular Weight : 434.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/zk-811752.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72364
Availability details for
Quantity
  • Availability
  • Add to Cart