ADVH49B83128 AChemBlock

MBX-8025 sodium salt

  • Molecular Weight : 466.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/mbx-8025-sodium-salt.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=K16161
Availability details for
Quantity
  • Availability
  • Add to Cart