ADVH49B83132 AChemBlock

7-(trifluoromethyl)chroman-4-one

  • Cas Number : 111141-02-7
  • Molecular Weight : 216.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/7-trifluoromethyl-chroman-4-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86876
Availability details for
Quantity
  • Availability
  • Add to Cart