ADVH49B83133 AChemBlock

(R)-1-(2,4-difluorophenyl)ethan-1-amine hydrochloride

  • Molecular Weight : 193.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/r-1-2-4-difluorophenyl-ethan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86879
Availability details for
Quantity
  • Availability
  • Add to Cart