ADVH49B8313C AChemBlock

(S)-1-(2,4-difluorophenyl)ethan-1-amine

  • Molecular Weight : 157.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/s-1-2-4-difluorophenyl-ethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86889
Availability details for
Quantity
  • Availability
  • Add to Cart