ADVH49B8313F AChemBlock

(S)-3,3-dimethylbutan-2-amine hydrochloride

  • Molecular Weight : 137.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/s-3-3-dimethylbutan-2-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86892
Availability details for
Quantity
  • Availability
  • Add to Cart