ADVH49B83148 AChemBlock

(R)-1-(3,5-difluorophenyl)ethan-1-amine

  • Cas Number : 771465-40-8
  • Molecular Weight : 157.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/r-1-3-5-difluorophenyl-ethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86917
Availability details for
Quantity
  • Availability
  • Add to Cart