ADVH49B83150 AChemBlock

(R)-2-((tert-butoxycarbonyl)amino)-3-(4-carbamoylphenyl)propanoic acid

  • Cas Number : 1213639-41-8
  • Molecular Weight : 308.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/r-2-tert-butoxycarbonyl-amino-3-4-carbamoylphenyl-propanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87223
Availability details for
Quantity
  • Availability
  • Add to Cart