ADVH49B83152 AChemBlock

(R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-carbamoylphenyl)propanoic acid

  • Cas Number : 1217610-39-3
  • Molecular Weight : 430.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/r-2-9h-fluoren-9-yl-methoxy-carbonyl-amino-3-4-carbamoylphenyl-propanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87225
Availability details for
Quantity
  • Availability
  • Add to Cart