ADVH49B83153 AChemBlock

(S)-3-(((benzyloxy)carbonyl)amino)-2-phenylpropanoic acid

  • Cas Number : 1233-79-0
  • Molecular Weight : 299.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/s-3-benzyloxy-carbonyl-amino-2-phenylpropanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87226
Availability details for
Quantity
  • Availability
  • Add to Cart