ADVH49B83155 AChemBlock

(S)-2-((tert-butoxycarbonyl)amino)-3-(4-carbamoylphenyl)propanoic acid

  • Cas Number : 205126-71-2
  • Molecular Weight : 308.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/s-2-tert-butoxycarbonyl-amino-3-4-carbamoylphenyl-propanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87234
Availability details for
Quantity
  • Availability
  • Add to Cart