ADVH49B83157 AChemBlock

(S)-2-amino-3-(4-carbamoylphenyl)propanoic acid

  • Cas Number : 223593-04-2
  • Molecular Weight : 208.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/s-2-amino-3-4-carbamoylphenyl-propanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87236
Availability details for
Quantity
  • Availability
  • Add to Cart