ADVH49B8315A AChemBlock

(S,E)-3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-6-(N'-((2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl)sulfonyl)formohydrazonamido)hexanoic acid

  • Cas Number : 401915-53-5
  • Molecular Weight : 662.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/s-e-3-9h-fluoren-9-yl-methoxy-carbonyl-amino-6-n-2-2-4-6-7-pentamethyl-2-3-dihydrobenzofuran-5-yl-sulfonyl-formohydrazonamido-hexanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87241
Availability details for
Quantity
  • Availability
  • Add to Cart