ADVH49B8315B AChemBlock

(S)-2-((((benzyloxy)carbonyl)amino)methyl)-3-methylbutanoic acid

  • Cas Number : 631899-14-4
  • Molecular Weight : 265.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/s-2-benzyloxy-carbonyl-amino-methyl-3-methylbutanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87242
Availability details for
Quantity
  • Availability
  • Add to Cart