ADVH49B83161 AChemBlock

(S)-2-(((tert-butoxycarbonyl)amino)methyl)-3,3-dimethylbutanoic acid

  • Molecular Weight : 245.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/s-2-tert-butoxycarbonyl-amino-methyl-3-3-dimethylbutanoic-acid-310558.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87249
Availability details for
Quantity
  • Availability
  • Add to Cart